accessibility menu, dialog, popup

UserName

CH3H3CCH3OONH2N

CAS RN 625471-18-3

IUPAC Name: tert-butyl 3-aminopiperidine-1-carboxylate
Synonyms: tert-butyl 3-aminopiperidine-1-carboxylate
Molecular Weight (g/mol): 200.28
Molecular Formula: C10H20N2O2
InChi Key: AKQXKEBCONUWCL-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1CCCC(N)C1
Percent Purity 
  • (1)
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)