missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-3-Amino-1-Boc-piperidine, 97%

Catalog No. AAH3053803
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3053803 1 g
AAH3053806 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3053803 Supplier Thermo Scientific Chemicals Supplier No. H3053803
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 625471-18-3 | C10H20N2O2 | 200.28 g/mol

(S)-(+)-3-Amino-1-Boc-piperidine is used to prepare selective noncovalent inhibitors of the bacterial cysteine protease IdeS. The product obtained by the reductive elimination reaction with ethylglyoxalte is used as a reference compound, which is used to determine the absolute configuration of the two enantiomers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(+)-3-Amino-1-Boc-piperidine is used to prepare selective noncovalent inhibitors of the bacterial cysteine protease IdeS. The product obtained by the reductive elimination reaction with ethylglyoxalte is used as a reference compound, which is used to determine the absolute configuration of the two enantiomers.

Solubility
Soluble in dimethylsulfoxide.

Notes
Air and carbon dioxide sensitive. Incompatible with strong oxidizing agents, strong acids and bases. Store in a cool place.

Chemical Identifiers

CAS 625471-18-3
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.28
MDL Number MFCD03094718
InChI Key AKQXKEBCONUWCL-UHFFFAOYNA-N
Synonym s-1-boc-3-aminopiperidine, s-3-amino-1-n-boc-piperidine, tert-butyl 3s-3-aminopiperidine-1-carboxylate, s-tert-butyl 3-aminopiperidine-1-carboxylate, s-3-amino-1-boc-piperidine, s-+-3-amino-1-boc-piperidine, 3s-3-aminopiperidine, n1-boc protected, s-1-boc-3-amniopiperidine, s-3-amino-piperidine-1-carboxylic acid tert-butyl ester, s-3-amino-n-tboc-piperidine
PubChem CID 1501975
IUPAC Name tert-butyl (3S)-3-aminopiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(N)C1

Specifications

Quantity 1 g
UN Number UN2735
Sensitivity Air Sensitive
Solubility Information Soluble in dimethylsulfoxide.
Formula Weight 200.28
Percent Purity 97%
Chemical Name or Material (S)-(+)-3-Amino-1-Boc-piperidine
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only