missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-3,3'-Dibromo-5,5',6,6',7,7',8,8'-octahydro-1,1'-bi-2-naphthol 97.0+%, TCI America™

Catalog No. D51071G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D51071G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D51071G Supplier TCI America Supplier No. D51071G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 765278-73-7
Molecular Formula C20H20Br2O2
Molecular Weight (g/mol) 452.186
MDL Number MFCD03093977
InChI Key WDRTXCNGVVLRSZ-UHFFFAOYSA-N
Synonym (S)-(-)-3,3′-Dibromo-5,5′,6,6′,7,7′,8,8′-octahydro-2,2′-dihydroxy-1,1′-binaphthyl
PubChem CID 3251420
IUPAC Name 3-bromo-1-(3-bromo-2-hydroxy-5,6,7,8-tetrahydronaphthalen-1-yl)-5,6,7,8-tetrahydronaphthalen-2-ol
SMILES C1CCC2=C(C(=C(C=C2C1)Br)O)C3=C4CCCCC4=CC(=C3O)Br

Specifications

Melting Point 144°C
Quantity 1 g
Formula Weight 452.19
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(-)-3,3′-Dibromo-5,5′,6,6′,7,7′,8,8′-octahydro-1,1′-bi-2-nap