missing translation for 'onlineSavingsMsg'
Learn More

(S)-3,3'-Dibromo-2,2'-dimethoxy-1,1'-binaphthyl 98.0+%, TCI America™

Catalog No. D2802200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D2802200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D2802200MG Supplier TCI America Supplier No. D2802200MG
Only null left

Chemical Identifiers

CAS 75714-60-2
Molecular Formula C22H16Br2O2
Molecular Weight (g/mol) 472.176
MDL Number MFCD04038415
InChI Key DFTUKDIMHCCQIT-UHFFFAOYSA-N
PubChem CID 394170
IUPAC Name 3-bromo-1-(3-bromo-2-methoxynaphthalen-1-yl)-2-methoxynaphthalene
SMILES COC1=C(C2=CC=CC=C2C=C1Br)C3=C(C(=CC4=CC=CC=C43)Br)OC

Specifications

Color White
Quantity 200 mg
Formula Weight 472.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-3,3′-Dibromo-2,2′-dimethoxy-1,1′-binaphthyl