accessibility menu, dialog, popup

UserName

H3COBrBrOH3C(?)

CAS RN 75714-60-2

IUPAC Name: 3,3'-dibromo-2,2'-dimethoxy-1,1'-binaphthalene
Synonyms: 3,3'-dibromo-2,2'-dimethoxy-1,1'-binaphthalene
Molecular Weight (g/mol): 472.18
Molecular Formula: C22H16Br2O2
InChi Key: DFTUKDIMHCCQIT-UHFFFAOYSA-N
SMILES: COC1=C(Br)C=C2C=CC=CC2=C1C1=C2C=CC=CC2=CC(Br)=C1OC
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results