missing translation for 'onlineSavingsMsg'
Learn More

(S)-3,3'-Dibromo-1,1'-bi-2-naphthol 97.0+%, TCI America™

Catalog No. p-7127515
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28111G 1 g
D28115G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D28111G Supplier TCI America Supplier No. D28111G

Chemical Identifiers

CAS 119707-74-3
Molecular Formula C20H12Br2O2
Molecular Weight (g/mol) 444.122
MDL Number MFCD03093629
InChI Key BRTBEAXHUYEXSY-UHFFFAOYSA-N
Synonym s---3,3'-dibromo-1,1'-bi-2-naphthol, r-3,3'-dibromo-1,1'-bi-2-naphthol, r-+-3,3'-dibromo-1,1'-bi-2-naphthol, s-3,3'-dibromo-1,1'-bi-2-naphthol, s-dibromo-1,1-binaphthol, r-dibromo-binol, s-dibromo-binol, r-dibromo-bi-2-naphthol, s-dibromo-bi-2-naphthol, r-dibromo-1,1-binaphthol
PubChem CID 10765693
IUPAC Name 3-bromo-1-(3-bromo-2-hydroxynaphthalen-1-yl)naphthalen-2-ol
SMILES C1=CC=C2C(=C1)C=C(C(=C2C3=C(C(=CC4=CC=CC=C43)Br)O)O)Br

Specifications

Quantity 1 g
Formula Weight 444.12
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-3,3′-Dibromo-1,1′-bi-2-naphthol