missing translation for 'onlineSavingsMsg'
Learn More

(S)-3-[1-(Dimethylamino)ethyl]phenol 98.0+%, TCI America™

Catalog No. D42585G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D42585G 5 g
D42581G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D42585G Supplier TCI America Supplier No. D42585G
Only null left

Chemical Identifiers

CAS 139306-10-8
Molecular Formula C10H15NO
Molecular Weight (g/mol) 165.24
MDL Number MFCD06656491
InChI Key GQZXRLWUYONVCP-UHFFFAOYNA-N
PubChem CID 445892
IUPAC Name 3-[1-(dimethylamino)ethyl]phenol
SMILES CC(N(C)C)C1=CC(O)=CC=C1

Specifications

Melting Point 119°C
Color White
Quantity 5 g
Formula Weight 165.24
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-3-[1-(Dimethylamino)ethyl]phenol