missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-3-[1-(Dimethylamino)ethyl]phenol 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 119°C |
| Color | White |
| Quantity | 1 g |
| Formula Weight | 165.24 |
| Percent Purity | ≥98.0% (GC,T) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | (S)-3-[1-(Dimethylamino)ethyl]phenol |
Safety and Handling
| TSCA | No |