Learn More
(S)-(+)-2-(Trifluoromethyl)pyrrolidine, 95%
CAS: 119580-41-5 | C5H8F3N | 139.121 g/mol
Supplier: Thermo Scientific Chemicals H2585403
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| (S)-(+)-2-(Trifluoromethyl)pyrrolidine | |
| 27°C to 31°C | |
| 26°C (79°F) | |
| MFCD03093766 | |
| UN2925 | |
| s-2-trifluoromethyl pyrrolidine, 2s-2-trifluoromethyl pyrrolidine, s-2-trifluoromethylpyrrolidine, 2 s-2-trifluoromethylpyrrolidine, s-+-2-trifluoromethyl pyrrolidine, s-2-trifluoromethyl-pyrrolidine, pyrrolidine, 2-trifluoromethyl-, s, 2s-2-trifluoromethylpyrrolidine, 2alpha-trifluoromethyl pyrrolidine, 2 s-2-trifluoromethyl pyrrolidine | |
| +10° (c=1 in Methanol) | |
| (2S)-2-(trifluoromethyl)pyrrolidine | |
| 2782840 | |
| 95% |
| 119580-41-5 | |
| 110°C to 112°C | |
| C5H8F3N | |
| 1 g | |
| Air Sensitive | |
| VINAMCOZNJHNIH-BYPYZUCNSA-N | |
| C1CC(NC1)C(F)(F)F | |
| 139.121 | |
| 139.12 |
Chemical Identifiers
| 119580-41-5 | |
| 139.121 | |
| VINAMCOZNJHNIH-BYPYZUCNSA-N | |
| (2S)-2-(trifluoromethyl)pyrrolidine |
| C5H8F3N | |
| MFCD03093766 | |
| 2782840 | |
| C1CC(NC1)C(F)(F)F |
Safety and Handling
GHS H Statement
H314-H318-H228
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.
P210-P240-P241-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H228-H314-H335
DOTInformation : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only