missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-(Trifluoromethyl)pyrrolidine, 95%

Catalog No. p-7028508
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH25854MD 250 mg
AAH2585403 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH25854MD Supplier Thermo Scientific Chemicals Supplier No. H25854MD
Only null left

CAS: 119580-41-5 | C5H8F3N | 139.121 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 119580-41-5
Molecular Formula C5H8F3N
Molecular Weight (g/mol) 139.121
MDL Number MFCD03093766
InChI Key VINAMCOZNJHNIH-BYPYZUCNSA-N
PubChem CID 2782840
IUPAC Name (2S)-2-(trifluoromethyl)pyrrolidine
SMILES C1CC(NC1)C(F)(F)F

Specifications

Melting Point 27°C to 31°C
Boiling Point 110°C to 112°C
Flash Point 26°C (79°F)
Quantity 250 mg
UN Number UN2925
Sensitivity Air Sensitive
Synonym s-2-trifluoromethyl pyrrolidine, 2s-2-trifluoromethyl pyrrolidine, s-2-trifluoromethylpyrrolidine, 2 s-2-trifluoromethylpyrrolidine, s-+-2-trifluoromethyl pyrrolidine, s-2-trifluoromethyl-pyrrolidine, pyrrolidine, 2-trifluoromethyl-, s, 2s-2-trifluoromethylpyrrolidine, 2alpha-trifluoromethyl pyrrolidine, 2 s-2-trifluoromethyl pyrrolidine
Optical Rotation +10° (c=1 in Methanol)
Formula Weight 139.12
Percent Purity 95%
Chemical Name or Material (S)-(+)-2-(Trifluoromethyl)pyrrolidine
Show More Show Less
Hazard Category H228-H314-H335
Hazard Statement GHS H Statement
H314-H318-H228
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable solid.
Precautionary Statement P210-P240-P241-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, CORROSIVE, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only