missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-(tert-Butoxycarbonyl)isoxazolidine-3-carboxylic Acid 98.0+%, TCI America™

Catalog No. B5176100MG
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5176100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B5176100MG Supplier TCI America Supplier No. B5176100MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1372202-46-4
Molecular Formula C9H15NO5
Molecular Weight (g/mol) 217.221
InChI Key FQNQHKBAZFREIU-LURJTMIESA-N
Synonym (S)-2-Boc-isoxazolidine-3-carboxylic Acid
PubChem CID 67558381
IUPAC Name (3S)-2-[(2-methylpropan-2-yl)oxycarbonyl]-1,2-oxazolidine-3-carboxylic acid
SMILES CC(C)(C)OC(=O)N1C(CCO1)C(=O)O

Specifications

Color White-Yellow
Quantity 100 mg
Formula Weight 217.22
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-2-(tert-Butoxycarbonyl)isoxazolidine-3-carboxylic Acid