accessibility menu, dialog, popup

UserName

CH3H3CCH3OONOOOH

CAS RN 1372202-46-4

IUPAC Name: 2-[(tert-butoxy)carbonyl]-1,2-oxazolidine-3-carboxylic acid
Synonyms: 2-(tert-butoxycarbonyl)-1,2-oxazolidine-3-carboxylic acid
Molecular Weight (g/mol): 217.22
Molecular Formula: C9H15NO5
InChi Key: FQNQHKBAZFREIU-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)N1OCCC1C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results