missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Phenylglycinol 97.0+%, TCI America™

Catalog No. p-7135060
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
5 g
Catalog No. Quantity
P12945G 5 g
P129425G 25 g
2 options

Catalog No. P12945G

Supplier: TCI America P12945G

Only null left

Chemical Identifiers

CAS 20989-17-7
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.182
MDL Number MFCD00064404
InChI Key IJXJGQCXFSSHNL-MRVPVSSYSA-N
Synonym s-+-2-phenylglycinol, s-2-phenylglycinol, s-2-amino-2-phenylethanol, s-phenylglycinol, h-phenylglycinol, 2s-2-amino-2-phenylethanol, 2s-2-amino-2-phenylethan-1-ol, l-2-phenylglycinol, h-phg-ol
PubChem CID 134797
IUPAC Name (2S)-2-amino-2-phenylethanol
SMILES C1=CC=C(C=C1)C(CO)N

Specifications

Melting Point 74°C
Color White-Yellow
Quantity 5 g
Formula Weight 137.18
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (S)-(+)-2-Phenylglycinol
TSCA No
Recommended Storage Refrigerator