accessibility menu, dialog, popup

UserName

CAS RN 20989-17-7

IUPAC Name: (2S)-2-amino-2-phenylethan-1-ol
Synonyms: (2S)-2-amino-2-phenylethanol
Molecular Weight (g/mol): 137.18
Molecular Formula: C8H11NO
InChi Key: IJXJGQCXFSSHNL-MRVPVSSYSA-N
SMILES: N[C@H](CO)C1=CC=CC=C1
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (6)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)