missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Phenylglycinol, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 307570250

Quantity 25g
Packaging Glass bottle

 View more versions of this product

Catalog No. AC307570250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

20989-17-7
137.182
IJXJGQCXFSSHNL-MRVPVSSYSA-N
134797
C1=CC=C(C=C1)C(CO)N
C8H11NO
MFCD00064404
s-+-2-phenylglycinol, s-2-phenylglycinol, s-2-amino-2-phenylethanol, s-phenylglycinol, h-phenylglycinol, 2s-2-amino-2-phenylethanol, 2s-2-amino-2-phenylethan-1-ol, l-2-phenylglycinol, h-phg-ol
(2S)-2-amino-2-phenylethanol

Specifications

(S)-(+)-2-Phenylglycinol
20989-17-7
97.5
+ 25.50
98%
C6H5CH(NH2)CH2OH
s-+-2-phenylglycinol, s-2-phenylglycinol, s-2-amino-2-phenylethanol, s-phenylglycinol, h-phenylglycinol, 2s-2-amino-2-phenylethanol, 2s-2-amino-2-phenylethan-1-ol, l-2-phenylglycinol, h-phg-ol
C1=CC=C(C=C1)C(CO)N
137.182
137.18
Authentic
Glass bottle
100.0
25g
C8H11NO
MFCD00064404
IJXJGQCXFSSHNL-MRVPVSSYSA-N
(2S)-2-amino-2-phenylethanol
134797
98%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents