Learn More
(S)-(+)-2-Phenylglycinol, 98+%
CAS: 20989-17-7 | C8H11NO | 137.182 g/mol
Supplier: Thermo Scientific Chemicals L1326506
Description
(S)-(+)-2-Phenylglycinol is used in the synthesis of chiral, unsymmetrical bisoxazolines. It is used as a reagent for the resolution of acids via the easily hydrolyzed amides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications(S)-(+)-2-Phenylglycinol is used in the synthesis of chiral, unsymmetrical bisoxazolines. It is used as a reagent for the resolution of acids via the easily hydrolyzed amides.
Solubility
Soluble in chloroform, dimethylsulfoxide and methanol. Limited solubility in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to air. Incompatible with oxidizing agents. Store at -20°C.
Specifications
| (S)-(+)-2-Phenylglycinol | |
| 76°C to 78°C | |
| MFCD00064404 | |
| 3196190 | |
| s-+-2-phenylglycinol, s-2-phenylglycinol, s-2-amino-2-phenylethanol, s-phenylglycinol, h-phenylglycinol, 2s-2-amino-2-phenylethanol, 2s-2-amino-2-phenylethan-1-ol, l-2-phenylglycinol, h-phg-ol | |
| IJXJGQCXFSSHNL-MRVPVSSYSA-N | |
| C1=CC=C(C=C1)C(CO)N | |
| 137.182 | |
| 137.18 |
| 20989-17-7 | |
| C8H11NO | |
| 5 g | |
| Air Sensitive | |
| Soluble in chloroform,dimethylsulfoxide and methanol. Limited solubility in water. | |
| +30° (c=0.75 in 1N HCl) | |
| (2S)-2-amino-2-phenylethanol | |
| 134797 | |
| ≥98% |
Chemical Identifiers
| 20989-17-7 | |
| 137.182 | |
| IJXJGQCXFSSHNL-MRVPVSSYSA-N | |
| 134797 | |
| C1=CC=C(C=C1)C(CO)N |
| C8H11NO | |
| MFCD00064404 | |
| s-+-2-phenylglycinol, s-2-phenylglycinol, s-2-amino-2-phenylethanol, s-phenylglycinol, h-phenylglycinol, 2s-2-amino-2-phenylethanol, 2s-2-amino-2-phenylethan-1-ol, l-2-phenylglycinol, h-phg-ol | |
| (2S)-2-amino-2-phenylethanol |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
DOTInformation : Hazard Class: 8; Packaging Group: III
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only