missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Phenylglycinol, 98+%

Catalog No. AAL1326503
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1326503 1 g
AAL1326506 5 g
AAL1326514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1326503 Supplier Thermo Scientific Chemicals Supplier No. L1326503

CAS: 20989-17-7 | C8H11NO | 137.182 g/mol

(S)-(+)-2-Phenylglycinol is used in the synthesis of chiral, unsymmetrical bisoxazolines. It is used as a reagent for the resolution of acids via the easily hydrolyzed amides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(S)-(+)-2-Phenylglycinol is used in the synthesis of chiral, unsymmetrical bisoxazolines. It is used as a reagent for the resolution of acids via the easily hydrolyzed amides.

Solubility
Soluble in chloroform, dimethylsulfoxide and methanol. Limited solubility in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. It is sensitive to air. Incompatible with oxidizing agents. Store at -20°C.

Chemical Identifiers

CAS 20989-17-7
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.182
MDL Number MFCD00064404
InChI Key IJXJGQCXFSSHNL-MRVPVSSYSA-N
Synonym s-+-2-phenylglycinol, s-2-phenylglycinol, s-2-amino-2-phenylethanol, s-phenylglycinol, h-phenylglycinol, 2s-2-amino-2-phenylethanol, 2s-2-amino-2-phenylethan-1-ol, l-2-phenylglycinol, h-phg-ol
PubChem CID 134797
IUPAC Name (2S)-2-amino-2-phenylethanol
SMILES C1=CC=C(C=C1)C(CO)N

Specifications

Melting Point 76°C to 78°C
Quantity 1 g
Beilstein 3196190
Sensitivity Air Sensitive
Solubility Information Soluble in chloroform,dimethylsulfoxide and methanol. Limited solubility in water.
Optical Rotation +30° (c=0.75 in 1N HCl)
Formula Weight 137.18
Percent Purity ≥98%
Chemical Name or Material (S)-(+)-2-Phenylglycinol
Hazard Category H314-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: III
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only