missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Phenylbutyric acid, 99%

Catalog No. AAL1275403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1275403 1 g
AAL1275406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1275403 Supplier Thermo Scientific Chemicals Supplier No. L1275403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 4286-15-1 | C10H12O2 | 164.204 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4286-15-1
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.204
MDL Number MFCD00063164
InChI Key OFJWFSNDPCAWDK-VIFPVBQESA-N
Synonym s-+-2-phenylbutyric acid, s-2-phenylbutanoic acid, 2s-2-phenylbutanoic acid, s-2-phenylbutyric acid, pubchem9874, 2-phenylbutyric acid, s, s--2-phenyl-mutyric acid, s-+-2-phenylbutanoic acid, s-+-2-phenyl-mutyric acid
PubChem CID 785329
IUPAC Name (2S)-2-phenylbutanoic acid
SMILES CCC(C1=CC=CC=C1)C(=O)O

Specifications

Density 1.055
Boiling Point 270°C to 272°C
Flash Point >110°C (230°F)
Refractive Index 1.515
Quantity 1 g
Beilstein 2045208
Optical Rotation +100° (c=5 in Toluene)
Formula Weight 164.2
Percent Purity 99%
Chemical Name or Material (S)-(+)-2-Phenylbutyric acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only