missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Pentanol, 97%

CAS: 26184-62-3 | C5H12O | 88.15 g/mol

Supplier:  Thermo Scientific Chemicals L09314MD

 View more versions of this product

Catalog No. AAL09314MD


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

(S)-(+)-2-Pentanol
0.81
33°C (91°F)
1.406
250 mg
1718820
JYVLIDXNZAXMDK-YFKPBYRVSA-N
CCCC(C)O
88.15
88.15
26184-62-3
118°C to 119°C
C5H12O
MFCD00065952
UN1105
s-+-2-pentanol, s-2-pentanol, 2s-pentan-2-ol, 2-pentanol, 2s, s---2-pentanol, 2s-2-pentanol, s +-2-pentanol
+14° (Neat)
(2S)-pentan-2-ol
2724896
97%

Chemical Identifiers

26184-62-3
88.15
JYVLIDXNZAXMDK-YFKPBYRVSA-N
2724896
CCCC(C)O
C5H12O
MFCD00065952
s-+-2-pentanol, s-2-pentanol, 2s-pentan-2-ol, 2-pentanol, 2s, s---2-pentanol, 2s-2-pentanol, s +-2-pentanol
(2S)-pentan-2-ol
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H226-H332-H335
Flammable liquid and vapor.
Harmful if inhaled.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P271-P280-P303+P361+P353-P304+P340-P312-P370+P378q-P501c

H226-H332-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: PENTANOLS

RTECSNumber : SA4900000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only