missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-Octyl 4-[4-(Hexyloxy)benzoyloxy]benzoate 98.0+%, TCI America™

Catalog No. O04611G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O04611G 1 g
O04615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O04611G Supplier TCI America Supplier No. O04611G

Chemical Identifiers

CAS 87321-20-8
Molecular Formula C28H38O5
Molecular Weight (g/mol) 454.607
MDL Number MFCD01941070
InChI Key PLGPDUBTEHIWRH-QFIPXVFZSA-N
Synonym 4-[4-(Hexyloxy)benzoyloxy]benzoic Acid (S)-2-Octyl Ester
PubChem CID 22845344
IUPAC Name [4-[(2S)-octan-2-yl]oxycarbonylphenyl] 4-hexoxybenzoate
SMILES CCCCCCC(C)OC(=O)C1=CC=C(C=C1)OC(=O)C2=CC=C(C=C2)OCCCCCC

Specifications

Melting Point 49°C
Color White
Quantity 1 g
Formula Weight 454.61
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material (S)-2-Octyl 4-[4-(Hexyloxy)benzoyloxy]benzoate