accessibility menu, dialog, popup

UserName

CAS RN 87321-20-8

IUPAC Name: octan-2-yl 4-[4-(hexyloxy)benzoyloxy]benzoate
Synonyms: octan-2-yl 4-[4-(hexyloxy)benzoyloxy]benzoate
Molecular Weight (g/mol): 454.61
Molecular Formula: C28H38O5
InChi Key: PLGPDUBTEHIWRH-UHFFFAOYNA-N
SMILES: CCCCCCOC1=CC=C(C=C1)C(=O)OC1=CC=C(C=C1)C(=O)OC(C)CCCCCC
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results