missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Octanol 98.0+%, TCI America™

Supplier:  TCI America O01445ML

 View more versions of this product

Catalog No. O01445ML


Only null left

Description

Description

Specifications

Specifications

(S)-(+)-2-Octanol
Colorless
MFCD00064283
s-+-2-octanol, s-octan-2-ol, s-2-octanol, 2s-octan-2-ol, d +-2-octanol, d-octan-2-ol, 2s-2-octanol, 2-octanol, 2s, l-2-octanol, 2-octanol s
CCCCCCC(C)O
130.23
CHEBI:37870
≥98.0% (GC)
6169-06-8
C8H18O
5 mL
SJWFXCIHNDVPSH-UHFFFAOYNA-N
(2S)-octan-2-ol
2723888
130.23
Liquid

Chemical Identifiers

6169-06-8
130.23
SJWFXCIHNDVPSH-UHFFFAOYNA-N
2723888
(2S)-octan-2-ol
C8H18O
MFCD00064283
s-+-2-octanol, s-octan-2-ol, s-2-octanol, 2s-octan-2-ol, d +-2-octanol, d-octan-2-ol, 2s-2-octanol, 2-octanol, 2s, l-2-octanol, 2-octanol s
CHEBI:37870
CCCCCCC(C)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0217

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions