Learn More
(S)-(+)-2-Methylbutyric acid, 98%
CAS: 1730-91-2 | C5H10O2 | 102.13 g/mol
$233.34 - $233.34
Chemical Identifiers
CAS | 1730-91-2 |
---|---|
Molecular Formula | C5H10O2 |
Molecular Weight (g/mol) | 102.13 |
MDL Number | MFCD00063166 |
InChI Key | WLAMNBDJUVNPJU-BYPYZUCNSA-N |
Synonym | s-+-2-methylbutyric acid, s-2-methylbutanoic acid, s-2-methylbutyric acid, 2s-2-methylbutanoic acid, 2-methylbutyric acid, s, s-+-2-methylbutanoic acid, smb, s-2-methylbutanoate, butanoic acid, 2-methyl-, s, +-2-methylbutanoic acid |
PubChem CID | 448893 |
ChEBI | CHEBI:38655 |
IUPAC Name | (2S)-2-methylbutanoic acid |
SMILES | CCC(C)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | |||||
AC224190010
|
Thermo Scientific Chemicals
224190010 |
1 g | Glass bottle |
Each for $233.34
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1730-91-2 | |
102.13 | |
WLAMNBDJUVNPJU-BYPYZUCNSA-N | |
448893 | |
(2S)-2-methylbutanoic acid |
C5H10O2 | |
MFCD00063166 | |
s-+-2-methylbutyric acid, s-2-methylbutanoic acid, s-2-methylbutyric acid, 2s-2-methylbutanoic acid, 2-methylbutyric acid, s, s-+-2-methylbutanoic acid, smb, s-2-methylbutanoate, butanoic acid, 2-methyl-, s, +-2-methylbutanoic acid | |
CHEBI:38655 | |
CCC(C)C(=O)O |
Specifications
1730-91-2 | |
100.0 | |
0.9380g/mL | |
73°C | |
97.5% min. (GC) | |
C5H10O2 | |
CH3CH2CH(CH3)COOH | |
1 g | |
0.938 | |
+ 19.00 | |
WLAMNBDJUVNPJU-BYPYZUCNSA-N | |
(2S)-2-methylbutanoic acid | |
448893 | |
102.13 | |
Liquid |
97.5 | |
Colorless | |
78.0°C (15.0 mmHg) | |
Authentic | |
Glass bottle | |
1.4041 to 1.4061 | |
MFCD00063166 | |
02, 304 | |
+19° (20°C c=neat) | |
s-+-2-methylbutyric acid, s-2-methylbutanoic acid, s-2-methylbutyric acid, 2s-2-methylbutanoic acid, 2-methylbutyric acid, s, s-+-2-methylbutanoic acid, smb, s-2-methylbutanoate, butanoic acid, 2-methyl-, s, +-2-methylbutanoic acid | |
CCC(C)C(=O)O | |
102.13 | |
CHEBI:38655 | |
98% | |
(S)-(+)-2-Methylbutyric acid |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water
GHS Signal Word: Danger
RUO – Research Use Only