missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Methylbutyric acid, 98%

CAS: 1730-91-2 | C5H10O2 | 102.13 g/mol

Supplier:  Thermo Scientific Chemicals 224190010

 View more versions of this product

Catalog No. AC224190010


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

(S)-(+)-2-Methylbutyric acid
97.5
Colorless
78.0°C (15.0 mmHg)
Authentic
Glass bottle
1.4041 to 1.4061
MFCD00063166
02, 304
+19° (20°C c=neat)
s-+-2-methylbutyric acid, s-2-methylbutanoic acid, s-2-methylbutyric acid, 2s-2-methylbutanoic acid, 2-methylbutyric acid, s, s-+-2-methylbutanoic acid, smb, s-2-methylbutanoate, butanoic acid, 2-methyl-, s, +-2-methylbutanoic acid
CCC(C)C(=O)O
102.13
CHEBI:38655
98%
1730-91-2
100.0
0.9380g/mL
73°C
97.5% min. (GC)
C5H10O2
CH3CH2CH(CH3)COOH
1 g
0.938
+ 19.00
WLAMNBDJUVNPJU-BYPYZUCNSA-N
(2S)-2-methylbutanoic acid
448893
102.13
Liquid

Chemical Identifiers

1730-91-2
102.13
WLAMNBDJUVNPJU-BYPYZUCNSA-N
448893
(2S)-2-methylbutanoic acid
C5H10O2
MFCD00063166
s-+-2-methylbutyric acid, s-2-methylbutanoic acid, s-2-methylbutyric acid, 2s-2-methylbutanoic acid, 2-methylbutyric acid, s, s-+-2-methylbutanoic acid, smb, s-2-methylbutanoate, butanoic acid, 2-methyl-, s, +-2-methylbutanoic acid
CHEBI:38655
CCC(C)C(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only