missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-Methylbutyl p-Toluenesulfonate 95.0+%, TCI America™

Catalog No. p-7136861
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T15955G 5 g
T159525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T15955G Supplier TCI America Supplier No. T15955G

Chemical Identifiers

CAS 38261-81-3
Molecular Formula C12H18O3S
Molecular Weight (g/mol) 242.33
MDL Number MFCD00013399
InChI Key HPEVJTNZYIMANV-UHFFFAOYNA-N
Synonym p-Toluenesulfonic Acid (S)-2-Methylbutyl Ester
PubChem CID 113324
IUPAC Name 2-methylbutyl 4-methylbenzene-1-sulfonate
SMILES CCC(C)COS(=O)(=O)C1=CC=C(C)C=C1

Specifications

Color Yellow
Quantity 5 g
Formula Weight 242.33
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-2-Methylbutyl p-Toluenesulfonate