accessibility menu, dialog, popup

UserName

CAS RN 38261-81-3

IUPAC Name: 2-methylbutyl 4-methylbenzene-1-sulfonate
Synonyms: 2-methylbutyl 4-methylbenzenesulfonate
Molecular Weight (g/mol): 242.33
Molecular Formula: C12H18O3S
InChi Key: HPEVJTNZYIMANV-UHFFFAOYNA-N
SMILES: CCC(C)COS(=O)(=O)C1=CC=C(C)C=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results