Learn More
(S)-(-)-2-Methyl-CBS-oxazaborolidine, 1M solution in toluene, Thermo Scientific Chemicals
$446.14 - $446.14
Chemical Identifiers
| CAS | 112022-81-8 |
|---|---|
| Molecular Formula | C18H20BNO |
| Molecular Weight (g/mol) | 277.17 |
| MDL Number | MFCD00078439 |
| InChI Key | VMKAFJQFKBASMU-KRWDZBQOSA-N |
| Synonym | s-2-methyl-cbs-oxazaborolidine, s-methyl oxazaborolidine, s---2-methyl-cbs-oxazaborolidine, s-5,5-diphenyl-2-methyl-3,4-propano-1,3,2-oxazaborolidine, corey catalyst, s-methyl-cbs-oxazaborolidine, s-3,3-diphenyl-1-methylpyrrolidino 1,2-c-1,3,2-oxazaborole, s-me-cbs catalyst, s-methyl-cbs, s-me-cbs |
| PubChem CID | 2734713 |
| IUPAC Name | (3aS)-1-methyl-3,3-diphenyl-3a,4,5,6-tetrahydropyrrolo[1,2-c][1,3,2]oxazaborole |
| SMILES | [H][C@@]12CCCN1B(C)OC2(C1=CC=CC=C1)C1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC369770250
|
Thermo Scientific Chemicals
369770250 |
25 mL | Glass bottle |
Each for $446.14
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 112022-81-8 | |
| 277.17 | |
| VMKAFJQFKBASMU-KRWDZBQOSA-N | |
| 2734713 | |
| [H][C@@]12CCCN1B(C)OC2(C1=CC=CC=C1)C1=CC=CC=C1 |
| C18H20BNO | |
| MFCD00078439 | |
| s-2-methyl-cbs-oxazaborolidine, s-methyl oxazaborolidine, s---2-methyl-cbs-oxazaborolidine, s-5,5-diphenyl-2-methyl-3,4-propano-1,3,2-oxazaborolidine, corey catalyst, s-methyl-cbs-oxazaborolidine, s-3,3-diphenyl-1-methylpyrrolidino 1,2-c-1,3,2-oxazaborole, s-me-cbs catalyst, s-methyl-cbs, s-me-cbs | |
| (3aS)-1-methyl-3,3-diphenyl-3a,4,5,6-tetrahydropyrrolo[1,2-c][1,3,2]oxazaborole |
Specifications
| 112022-81-8,108-88-3 | |
| 32.82,71.66 | |
| 111.0°C | |
| Glass bottle | |
| MFCD00078439 | |
| 0.929 | |
| Solubility in water: reacts | |
| [H][C@@]12CCCN1B(C)OC2(C1=CC=CC=C1)C1=CC=CC=C1 | |
| 277.17 | |
| 277.17 |
| 28.34,67.18 | |
| 0.9290g/mL | |
| 4°C | |
| C18H20BNO | |
| 25 mL | |
| s-2-methyl-cbs-oxazaborolidine, s-methyl oxazaborolidine, s---2-methyl-cbs-oxazaborolidine, s-5,5-diphenyl-2-methyl-3,4-propano-1,3,2-oxazaborolidine, corey catalyst, s-methyl-cbs-oxazaborolidine, s-3,3-diphenyl-1-methylpyrrolidino 1,2-c-1,3,2-oxazaborole, s-me-cbs catalyst, s-methyl-cbs, s-me-cbs | |
| VMKAFJQFKBASMU-KRWDZBQOSA-N | |
| (3aS)-1-methyl-3,3-diphenyl-3a,4,5,6-tetrahydropyrrolo[1,2-c][1,3,2]oxazaborole | |
| 2734713 | |
| (S)-(-)-2-Methyl-CBS-oxazaborolidine, 1M solution in toluene |
Safety and Handling
GHS H Statement
Harmful if swallowed.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Causes serious eye damage.
Suspected of damaging the unborn child.
May cause damage to organs through prolong
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF INHALED: Remov
GHS Signal Word: Danger
RUO – Research Use Only