missing translation for 'onlineSavingsMsg'
Learn More

S-(2-Methyl-3-furyl) Thioacetate 96.0+%, TCI America™

Catalog No. M26615G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M26615G 5 g
M266125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M26615G Supplier TCI America Supplier No. M26615G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 55764-25-5
Molecular Formula C7H8O2S
Molecular Weight (g/mol) 156.20
MDL Number MFCD01632595
InChI Key PQFIBPDAGFGLBY-UHFFFAOYSA-N
Synonym 2-methylfuran-3-thiol acetate, 3-acetylthio-2-methylfuran, ethanethioic acid, s-2-methyl-3-furanyl ester, s-2-methyl-3-furyl thioacetate, s-2-methyl-3-furyl ethanethioate, 2-methyl-3-furanthiol, acetate, unii-7277jee7af, s-2-methylfuran-3-yl ethanethioate, thioacetic acid s-2-methyl-3-furyl ester, 1-2-methylfuran-3-yl sulfanyl ethanone
PubChem CID 108765
IUPAC Name 1-[(2-methylfuran-3-yl)sulfanyl]ethan-1-one
SMILES CC(=O)SC1=C(C)OC=C1

Specifications

Boiling Point 235°C
Quantity 5 g
UN Number 3272
Formula Weight 156.20
Percent Purity ≥96.0% (GC)
Physical Form Liquid
Chemical Name or Material S-(2-Methyl-3-furyl) Thioacetate