accessibility menu, dialog, popup

UserName

CH3OSOH3C

CAS RN 55764-25-5

IUPAC Name: 1-[(2-methylfuran-3-yl)sulfanyl]ethan-1-one
Synonyms: 1-[(2-methylfuran-3-yl)sulfanyl]ethanone
Molecular Weight (g/mol): 156.2
Molecular Formula: C7H8O2S
InChi Key: PQFIBPDAGFGLBY-UHFFFAOYSA-N
SMILES: CC(=O)SC1=C(C)OC=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results