missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-2-Methoxypropionic acid, 98%

Catalog No. p-158292
Change view
Click to view available options
Quantity:
500 mg
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC331725000 500 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC331725000 Supplier Thermo Scientific Chemicals Supplier No. 331725000

CAS: 23953-00-6 | C4H8O3 | 104.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 23953-00-6
Molecular Formula C4H8O3
Molecular Weight (g/mol) 104.11
MDL Number MFCD01632589
InChI Key ICPWFHKNYYRBSZ-VKHMYHEASA-N
Synonym s-2-methoxypropanoic acid, s---2-methoxypropionic acid, 2s-2-methoxypropanoic acid, propanoic acid, 2-methoxy-, 2s, unii-74k293ocx6, 2-methoxypropanoic acid, s, propanoic acid,2-methoxy-, 2s, propanoic acid, 2-methoxy-, s, propionic acid, 2-methoxy-, s--, s-2-methoxypropionic acid
PubChem CID 5702566
IUPAC Name (2S)-2-methoxypropanoic acid
SMILES CC(C(=O)O)OC

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Boiling Point 99.0°C (20.0 mmHg)
Flash Point 87°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4132 to 1.4152
Linear Formula CH3CH(OCH3)COOH
Quantity 500 mg
Beilstein 03, III, 444
Specific Rotation Condition −75.00 (20.00°C c=neat)
Specific Rotation − 75.00
Formula Weight 104.11
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material (S)-(-)-2-Methoxypropionic acid
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
TSCA TSCA

RUO – Research Use Only