accessibility menu, dialog, popup

UserName

CH3O(S)CH3OO

CAS RN 23953-00-6

IUPAC Name: (2S)-2-methoxypropanoate
Synonyms: (2S)-2-methoxypropanoate
Molecular Weight (g/mol): 103.1
Molecular Formula: C4H7O3
InChi Key: ICPWFHKNYYRBSZ-VKHMYHEASA-M
SMILES: CO[C@@H](C)C([O-])=O
Boiling Point 
  • (2)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)