missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-2-(Methoxymethyl)pyrrolidine 98.0+%, TCI America™
Supplier: TCI America M11615G
Specifications
| (S)-2-(Methoxymethyl)pyrrolidine | |
| Yellow | |
| C6H13NO | |
| 5 g | |
| s-2-methoxymethyl pyrrolidine, s-+-2-methoxymethyl pyrrolidine, 2s-2-methoxymethyl pyrrolidine, o-methyl-l-prolinol, s-2-methoxymethyl-pyrrolidine, s-2-methoxymethylpyrrolidine, s-+-2-methoxymethyl pyrolidine, pyrrolidine, 2-methoxymethyl-, 2s, s-+-2-methoxymethyl pyrrplidine, 2s pyrrolidin-2-yl methoxymethane | |
| COCC1CCCN1 | |
| 115.176 | |
| 115.18 | |
| Liquid |
| 63126-47-6 | |
| 68°C | |
| MFCD00010408 | |
| 1993 | |
| CHPRFKYDQRKRRK-LURJTMIESA-N | |
| (2S)-2-(methoxymethyl)pyrrolidine | |
| 671217 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 63126-47-6 | |
| 115.176 | |
| CHPRFKYDQRKRRK-LURJTMIESA-N | |
| 671217 | |
| COCC1CCCN1 |
| C6H13NO | |
| MFCD00010408 | |
| s-2-methoxymethyl pyrrolidine, s-+-2-methoxymethyl pyrrolidine, 2s-2-methoxymethyl pyrrolidine, o-methyl-l-prolinol, s-2-methoxymethyl-pyrrolidine, s-2-methoxymethylpyrrolidine, s-+-2-methoxymethyl pyrolidine, pyrrolidine, 2-methoxymethyl-, 2s, s-+-2-methoxymethyl pyrrplidine, 2s pyrrolidin-2-yl methoxymethane | |
| (2S)-2-(methoxymethyl)pyrrolidine |
Safety and Handling
TSCA : No