missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-(Methoxymethyl)pyrrolidine 98.0+%, TCI America™

Catalog No. p-7132569
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M11611G 1 g
M11615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M11611G Supplier TCI America Supplier No. M11611G

Chemical Identifiers

CAS 63126-47-6
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.176
MDL Number MFCD00010408
InChI Key CHPRFKYDQRKRRK-LURJTMIESA-N
Synonym s-2-methoxymethyl pyrrolidine, s-+-2-methoxymethyl pyrrolidine, 2s-2-methoxymethyl pyrrolidine, o-methyl-l-prolinol, s-2-methoxymethyl-pyrrolidine, s-2-methoxymethylpyrrolidine, s-+-2-methoxymethyl pyrolidine, pyrrolidine, 2-methoxymethyl-, 2s, s-+-2-methoxymethyl pyrrplidine, 2s pyrrolidin-2-yl methoxymethane
PubChem CID 671217
IUPAC Name (2S)-2-(methoxymethyl)pyrrolidine
SMILES COCC1CCCN1

Specifications

Color Yellow
Boiling Point 68°C
Quantity 1 g
UN Number 1993
Formula Weight 115.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material (S)-2-(Methoxymethyl)pyrrolidine