missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-Diphenylmethylpyrrolidine, 97+%

Catalog No. p-3757780
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC348372500 250 mg Glass bottle
AC348370010 1 g Glass bottle
AC348370050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC348372500 Supplier Thermo Scientific Chemicals Supplier No. 348372500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 119237-64-8 | C17H20N | 238.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 119237-64-8
Molecular Formula C17H20N
Molecular Weight (g/mol) 238.35
MDL Number MFCD00799525
InChI Key OXOBKZZXZVFOBB-INIZCTEOSA-O
Synonym s-2-benzhydrylpyrrolidine, s---2-diphenylmethyl pyrrolidine, s-2-diphenylmethylpyrrolidine, 2s-2-diphenylmethyl pyrrolidine, 2s-2-benzhydrylpyrrolidine, 2s-2-benzylhydrylpyrrolidine, 2r-2-benzhydrylpyrrolidine, pubchem18168, 2alpha-benzhydrylpyrrolidine, s-2-benzhydryl-pyrrolidine
PubChem CID 2734053
SMILES C1C[NH2+][C@@H](C1)C(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

CAS Min % 97.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0620g/mL
Flash Point 109°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Refractive Index 1.5860 to 1.5880
Quantity 250 mg
Specific Gravity 1.062
Specific Rotation Condition - 3.00 (25.00°C C=1,Chloroform)
Specific Rotation − 3.00
IUPAC Name (2S)-2-benzhydrylpyrrolidine
Formula Weight 237.34
Percent Purity 97+%
Physical Form Oil
Chemical Name or Material (S)-2-Diphenylmethylpyrrolidine
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only