missing translation for 'onlineSavingsMsg'
Learn More

S-[2-(Dimethylamino)ethyl]isothiourea Dihydrochloride 97.0+%, TCI America™

Catalog No. D065210G
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D065210G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D065210G Supplier TCI America Supplier No. D065210G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 16111-27-6
Melting Point 181°C
Color White
Molecular Formula C5H13ClN3S
Quantity 10 g
MDL Number MFCD00060161
InChI Key NEQDKQTXUQVFFU-UHFFFAOYSA-N
SMILES [Cl].CN(C)CCSC(N)=N
IUPAC Name [2-(carbamimidoylsulfanyl)ethyl]dimethylamine hydrochloridyl
Molecular Weight (g/mol) 182.69
PubChem CID 85295
Formula Weight 220.16
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material S-[2-(Dimethylamino)ethyl]isothiourea Dihydrochloride
Show More Show Less
EINECSNumber (2)-3142
RTECSNumber UM3770000
TSCA Yes