missing translation for 'onlineSavingsMsg'
Learn More

(S)-(+)-2-Dibenzylamino-3-phenyl-1-propanol, 99%, Thermo Scientific Chemicals

Catalog No. p-7046309
Change view
Click to view available options
Quantity:
5 g
Catalog No. Quantity
AAL1391606 5 g
1 options

Catalog No. AAL1391606

Supplier: Thermo Scientific Chemicals L1391606

Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 111060-52-7
Molecular Formula C23H25NO
Molecular Weight (g/mol) 331.46
MDL Number MFCD00191984
InChI Key ZXNVOFMPUPOZDF-UHFFFAOYNA-N
Synonym s-+-2-dibenzylamino-3-phenyl-1-propanol, s-2-dibenzylamino-3-phenylpropan-1-ol, 2s-2-dibenzylamino-3-phenylpropan-1-ol, dibenzyl phenylalaninol, n,n-dibenzyl-l-phenylalaninol, 2-s-dibenzylamino-3-phenylpropan-1-ol, s-beta-dibenzylamino benzene-1-propanol, s--2-dibenzylamino-3-phenyl-1-propanol, benzenepropanol,b-bis phenylmethyl amino-, bs
PubChem CID 7019180
IUPAC Name (2S)-2-(dibenzylamino)-3-phenylpropan-1-ol
SMILES OCC(CC1=CC=CC=C1)N(CC1=CC=CC=C1)CC1=CC=CC=C1

Specifications

Melting Point 72°C to 74°C
Quantity 5 g
Beilstein 7253058
Optical Rotation +8.5° (c=4 in Methanol)
Formula Weight 331.46
Percent Purity 99%
Chemical Name or Material (S)-(+)-2-Dibenzylamino-3-phenyl-1-propanol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only