missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-2-Chloropropionic acid, 98%

CAS: 29617-66-1 | C3H4ClO2 | 107.51 g/mol

$320.12 - $320.12

Chemical Identifiers

CAS 29617-66-1
Molecular Formula C3H4ClO2
Molecular Weight (g/mol) 107.51
MDL Number MFCD00064205
InChI Key GAWAYYRQGQZKCR-REOHCLBHSA-M
Synonym s---2-chloropropionic acid, s-2-chloropropanoic acid, s-2-chloropropionic acid, 2s-2-chloropropanoic acid, l-2-chloropropanoic acid, l-2-chloropropionic acid, alpha-l-chloropropionic acid, unii-pcc7j322cm, 2s-chloropropanoic acid, 2 s-chloropropionic acid
PubChem CID 107915
ChEBI CHEBI:73956
IUPAC Name (2S)-2-chloropropanoic acid
SMILES C[C@H](Cl)C([O-])=O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC254970050
SDS View Documents
Thermo Scientific Chemicals
254970050
5 g Glass bottle
Each for $320.12
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

29617-66-1
107.51
GAWAYYRQGQZKCR-REOHCLBHSA-M
107915
(2S)-2-chloropropanoic acid
C3H4ClO2
MFCD00064205
s---2-chloropropionic acid, s-2-chloropropanoic acid, s-2-chloropropionic acid, 2s-2-chloropropanoic acid, l-2-chloropropanoic acid, l-2-chloropropionic acid, alpha-l-chloropropionic acid, unii-pcc7j322cm, 2s-chloropropanoic acid, 2 s-chloropropionic acid
CHEBI:73956
C[C@H](Cl)C([O-])=O

Specifications

29617-66-1
100.0
Yellow
187°C
Authentic
Glass bottle
1.4340 to 1.436
5 g
-14.5
Solubility in water: soluble in water. Other solubilities: soluble in ethanol and ether
C[C@H](Cl)C([O-])=O
107.51
CHEBI:73956
98%
(S)-(-)-2-Chloropropionic acid, 98%
97.5
4°C
1.2400g/mL
107°C
97.5% min. (GC)
C3H4ClO2
MFCD00064205
1.24
s---2-chloropropionic acid, s-2-chloropropanoic acid, s-2-chloropropionic acid, 2s-2-chloropropanoic acid, l-2-chloropropanoic acid, l-2-chloropropionic acid, alpha-l-chloropropionic acid, unii-pcc7j322cm, 2s-chloropropanoic acid, 2 s-chloropropionic acid
GAWAYYRQGQZKCR-REOHCLBHSA-M
(2S)-2-chloropropanoic acid
107915
108.52
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful in contact with skin.
Harmful if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

RTECSNumber : UA2451950

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only