missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-2-Chlorobutyric Acid 98.0+%, TCI America™
$258.21 - $888.56
Chemical Identifiers
| CAS | 32653-32-0 |
|---|---|
| Molecular Formula | C4H7ClO2 |
| Molecular Weight (g/mol) | 122.548 |
| MDL Number | MFCD00067099 |
| InChI Key | RVBUZBPJAGZHSQ-VKHMYHEASA-N |
| PubChem CID | 6999872 |
| IUPAC Name | (2S)-2-chlorobutanoic acid |
| SMILES | CCC(C(=O)O)Cl |
Chemical Identifiers
| 32653-32-0 | |
| 122.548 | |
| RVBUZBPJAGZHSQ-VKHMYHEASA-N | |
| (2S)-2-chlorobutanoic acid |
| C4H7ClO2 | |
| MFCD00067099 | |
| 6999872 | |
| CCC(C(=O)O)Cl |
Specifications
| 32653-32-0 | |
| 94°C | |
| MFCD00067099 | |
| 3265 | |
| CCC(C(=O)O)Cl | |
| 122.548 | |
| 122.55 | |
| Liquid |
| Yellow | |
| C4H7ClO2 | |
| 1 g | |
| RVBUZBPJAGZHSQ-VKHMYHEASA-N | |
| (2S)-2-chlorobutanoic acid | |
| 6999872 | |
| ≥98.0% (T) | |
| (S)-2-Chlorobutyric Acid |
Safety and Handling
TSCA : No