accessibility menu, dialog, popup

UserName

H3COOHCl

CAS RN 32653-32-0

IUPAC Name: 2-chlorobutanoic acid
Synonyms: 2-chlorobutanoic acid
Molecular Weight (g/mol): 122.55
Molecular Formula: C4H7ClO2
InChi Key: RVBUZBPJAGZHSQ-UHFFFAOYNA-N
SMILES: CCC(Cl)C(O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results