missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(S)-2-Chlorobutyric Acid 98.0+%, TCI America™
Specifications
Specifications
| Color | Yellow |
| Boiling Point | 94°C |
| Quantity | 5 g |
| UN Number | 3265 |
| Formula Weight | 122.55 |
| Percent Purity | ≥98.0% (T) |
| Physical Form | Liquid |
| Chemical Name or Material | (S)-2-Chlorobutyric Acid |
Safety and Handling
| TSCA | No |