missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-Chloro-4-methylvaleric Acid 97.0+%, TCI America™

Catalog No. C13715G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C13715G 5 g
C13711G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C13715G Supplier TCI America Supplier No. C13715G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 28659-81-6
Molecular Formula C6H11ClO2
Molecular Weight (g/mol) 150.602
MDL Number MFCD00044943
InChI Key CBQBIPRPIHIKPW-YFKPBYRVSA-N
Synonym (S)-2-Chloro-4-methylpentanoic Acid
PubChem CID 12732683
IUPAC Name (2S)-2-chloro-4-methylpentanoic acid
SMILES CC(C)CC(C(=O)O)Cl

Specifications

Color Yellow
Boiling Point 126°C
Quantity 5 g
Formula Weight 150.60
Percent Purity ≥97.0% (T)
Physical Form Liquid
Chemical Name or Material (S)-2-Chloro-4-methylvaleric Acid