accessibility menu, dialog, popup

UserName

CAS RN 28659-81-6

IUPAC Name: 2-chloro-4-methylpentanoic acid
Synonyms: 2-chloro-4-methylpentanoic acid
Molecular Weight (g/mol): 150.6
Molecular Formula: C6H11ClO2
InChi Key: CBQBIPRPIHIKPW-UHFFFAOYNA-N
SMILES: CC(C)CC(Cl)C(O)=O
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results