missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-(Boc-amino)-4-(Fmoc-amino)butyric acid, 95%

Catalog No. AAH5198903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5198903 1 g
AAH5198906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5198903 Supplier Thermo Scientific Chemicals Supplier No. H5198903
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 117106-21-5 | C24H28N2O6 | 440.496 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 117106-21-5
Molecular Formula C24H28N2O6
Molecular Weight (g/mol) 440.496
MDL Number MFCD00236845
InChI Key MJZDTTZGQUEOBL-FQEVSTJZSA-N
Synonym boc-dab fmoc-oh, s-4-9h-fluoren-9-yl methoxy carbonyl amino-2-tert-butoxycarbonyl amino butanoic acid, n-a-boc-n-g-fmoc-l-2,4-diaminobutyric acid, s-n2-boc-n4-fmoc-2,4-diaminobutyric acid, ambotzbaa1086, boc-dbu gamma-fmoc, boc-l-dab fmoc-oh, boc-l-2,4-diaminobutyric acid fmoc, n-boc-n'-fmoc-l-2,4-diaminobutyric acid, boc-dab fmoc-oh hplc
PubChem CID 2755945
IUPAC Name (2S)-4-(9H-fluoren-9-ylmethoxycarbonylamino)-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
SMILES CC(C)(C)OC(=O)NC(CCNC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13)C(=O)O

Specifications

Quantity 1 g
Beilstein 4771583
Formula Weight 440.5
Percent Purity 95%
Chemical Name or Material (S)-2-(Boc-amino)-4-(Fmoc-amino)butyric acid
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only