Learn More
(S)-(-)-2-(BOC-amino)-1,4-butanediol, 97%
CAS: 128427-10-1 | C9H19NO4 | 205.25 g/mol
$475.85 - $475.85
Chemical Identifiers
| CAS | 128427-10-1 |
|---|---|
| Molecular Formula | C9H19NO4 |
| Molecular Weight (g/mol) | 205.25 |
| MDL Number | MFCD03093946 |
| InChI Key | KLRRFBSWOIUAHZ-ZETCQYMHSA-N |
| Synonym | s---2-boc-amino-1,4-butanediol, s-n-boc-2-aminobutane-1,4-diol, s-n-boc-2-amino-butane-1,4-diol, s-tert-butyl 1,4-dihydroxybutan-2-ylcarbamate, s-tert-butyl 1,4-dihydroxybutan-2-yl carbamate, tert-butyl n-2s-1,4-dihydroxybutan-2-yl carbamate, s-2-tert-butoxycarbonylamino-1,4-butanediol, carbamic acid, 1s-3-hydroxy-1-hydroxymethyl propyl-, 1,1-dimethylethyl, pubchem10472, ksc496o7n |
| PubChem CID | 2734850 |
| IUPAC Name | tert-butyl N-[(2S)-1,4-dihydroxybutan-2-yl]carbamate |
| SMILES | CC(C)(C)OC(=O)N[C@H](CO)CCO |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC375100050
|
Thermo Scientific Chemicals
375100050 |
5 g |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 128427-10-1 | |
| 205.25 | |
| KLRRFBSWOIUAHZ-ZETCQYMHSA-N | |
| 2734850 | |
| CC(C)(C)OC(=O)N[C@H](CO)CCO |
| C9H19NO4 | |
| MFCD03093946 | |
| s---2-boc-amino-1,4-butanediol, s-n-boc-2-aminobutane-1,4-diol, s-n-boc-2-amino-butane-1,4-diol, s-tert-butyl 1,4-dihydroxybutan-2-ylcarbamate, s-tert-butyl 1,4-dihydroxybutan-2-yl carbamate, tert-butyl n-2s-1,4-dihydroxybutan-2-yl carbamate, s-2-tert-butoxycarbonylamino-1,4-butanediol, carbamic acid, 1s-3-hydroxy-1-hydroxymethyl propyl-, 1,1-dimethylethyl, pubchem10472, ksc496o7n | |
| tert-butyl N-[(2S)-1,4-dihydroxybutan-2-yl]carbamate |
Specifications
| 65.0°C to 69.0°C | |
| 96.0 | |
| White | |
| 96% min. (GC) | |
| Glass Bottle | |
| MFCD03093946 | |
| s---2-boc-amino-1,4-butanediol, s-n-boc-2-aminobutane-1,4-diol, s-n-boc-2-amino-butane-1,4-diol, s-tert-butyl 1,4-dihydroxybutan-2-ylcarbamate, s-tert-butyl 1,4-dihydroxybutan-2-yl carbamate, tert-butyl n-2s-1,4-dihydroxybutan-2-yl carbamate, s-2-tert-butoxycarbonylamino-1,4-butanediol, carbamic acid, 1s-3-hydroxy-1-hydroxymethyl propyl-, 1,1-dimethylethyl, pubchem10472, ksc496o7n | |
| CC(C)(C)OC(=O)N[C@H](CO)CCO | |
| 205.25 | |
| 2734850 | |
| 97% | |
| (S)-(-)-2-(BOC-amino)-1, 4-butanediol |
| 128427-10-1 | |
| 100.0 | |
| Authentic | |
| C9H19NO4 | |
| HOCH2CH2CH(CH2OH)NHCO2C(CH3)3 | |
| -7° to -9° (20°C, 589nm) (c=1, chloroform) | |
| KLRRFBSWOIUAHZ-ZETCQYMHSA-N | |
| tert-butyl N-[(2S)-1,4-dihydroxybutan-2-yl]carbamate | |
| 5 g | |
| 205.25 | |
| Crystalline Powder |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning
RUO – Research Use Only