accessibility menu, dialog, popup

UserName

CAS RN 128427-10-1

IUPAC Name: tert-butyl N-[(2S)-1,4-dihydroxybutan-2-yl]carbamate
Synonyms: tert-butyl N-[(2S)-1,4-dihydroxybutan-2-yl]carbamate
Molecular Weight (g/mol): 205.25
Molecular Formula: C9H19NO4
InChi Key: KLRRFBSWOIUAHZ-ZETCQYMHSA-N
SMILES: CC(C)(C)OC(=O)N[C@H](CO)CCO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results