missing translation for 'onlineSavingsMsg'
Learn More

(S)-(-)-2-(BOC-amino)-1,4-butanediol, 97%

Catalog No. AC375100050
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC375100050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC375100050 Supplier Thermo Scientific Chemicals Supplier No. 375100050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 128427-10-1 | C9H19NO4 | 205.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 128427-10-1
Molecular Formula C9H19NO4
Molecular Weight (g/mol) 205.25
MDL Number MFCD03093946
InChI Key KLRRFBSWOIUAHZ-ZETCQYMHSA-N
Synonym s---2-boc-amino-1,4-butanediol, s-n-boc-2-aminobutane-1,4-diol, s-n-boc-2-amino-butane-1,4-diol, s-tert-butyl 1,4-dihydroxybutan-2-ylcarbamate, s-tert-butyl 1,4-dihydroxybutan-2-yl carbamate, tert-butyl n-2s-1,4-dihydroxybutan-2-yl carbamate, s-2-tert-butoxycarbonylamino-1,4-butanediol, carbamic acid, 1s-3-hydroxy-1-hydroxymethyl propyl-, 1,1-dimethylethyl, pubchem10472, ksc496o7n
PubChem CID 2734850
IUPAC Name tert-butyl N-[(2S)-1,4-dihydroxybutan-2-yl]carbamate
SMILES CC(C)(C)OC(=O)N[C@H](CO)CCO

Specifications

Melting Point 65.0°C to 69.0°C
CAS Min % 96.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Linear Formula HOCH2CH2CH(CH2OH)NHCO2C(CH3)3
Specific Rotation -7° to -9° (20°C, 589nm) (c=1, chloroform)
Quantity 5 g
Formula Weight 205.25
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material (S)-(-)-2-(BOC-amino)-1, 4-butanediol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee

RUO – Research Use Only