missing translation for 'onlineSavingsMsg'
Learn More

(S)-2-(Benzyloxycarbonylamino)butyric acid, 98%, Thermo Scientific Chemicals

Catalog No. AAH6242614
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6242614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH6242614 Supplier Thermo Scientific Chemicals Supplier No. H6242614
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 42918-86-5
Molecular Formula C12H15NO4
Molecular Weight (g/mol) 237.255
MDL Number MFCD00077006
InChI Key SZQMTCSQWUYUML-JTQLQIEISA-N
Synonym z-abu-oh, s-2-benzyloxy carbonyl amino butanoic acid, 2s-2-benzyloxy carbonyl amino butanoic acid, z-2-abu-oh, s-2-bezyloxycarbonylamino butanoic acid, s-2-benzyloxycarbonylamino butanoic acid, butanoic acid, 2-phenylmethoxy carbonyl amino-, 2s, ambotzzaa1200, pubchem18943, s-2-cbz-amino butyricacid
PubChem CID 7349998
IUPAC Name (2S)-2-(phenylmethoxycarbonylamino)butanoic acid
SMILES CCC(C(=O)O)NC(=O)OCC1=CC=CC=C1

Specifications

Quantity 25 g
Formula Weight 237.25
Percent Purity 98%
Chemical Name or Material (S)-2-(Benzyloxycarbonylamino)butyric acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only