CAS RN 42918-86-5
IUPAC Name:
2-{[(benzyloxy)carbonyl]amino}butanoic acid
Synonyms:
2-{[(benzyloxy)carbonyl]amino}butanoic acid
Molecular Weight (g/mol):
237.26
Molecular Formula:
C12H15NO4
InChi Key:
SZQMTCSQWUYUML-UHFFFAOYNA-N
SMILES:
CCC(NC(=O)OCC1=CC=CC=C1)C(O)=O
1
–
1
of
1
results
1
Promotions Available
(S)-2-(Benzyloxycarbonylamino)butyric acid, 98%, Thermo Scientific Chemicals
(S)-2-(Benzyloxycarbonylamino)butyric acid, 98%, Thermo Scientific Chemicals
CAS: 42918-86-5 Molecular Formula: C12H15NO4 Molecular Weight (g/mol): 237.255 MDL Number: MFCD00077006 InChI Key: SZQMTCSQWUYUML-JTQLQIEISA-N Synonym: z-abu-oh,s-2-benzyloxy carbonyl amino butanoic acid,2s-2-benzyloxy carbonyl amino butanoic acid,z-2-abu-oh,s-2-bezyloxycarbonylamino butanoic acid,s-2-benzyloxycarbonylamino butanoic acid,butanoic acid, 2-phenylmethoxy carbonyl amino-, 2s,ambotzzaa1200,pubchem18943,s-2-cbz-amino butyricacid PubChem CID: 7349998 IUPAC Name: (2S)-2-(phenylmethoxycarbonylamino)butanoic acid SMILES: CCC(C(=O)O)NC(=O)OCC1=CC=CC=C1