missing translation for 'onlineSavingsMsg'
Learn More

S-(2-Benzothiazolyl) (Z)-2-(2-Amino-4-thiazolyl)-2-(methoxyimino)thioacetate 97.0+%, TCI America™

Catalog No. b36025g
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36025G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B36025G Supplier TCI America Supplier No. B36025G
Only null left

Chemical Identifiers

CAS 80756-85-0
Molecular Formula C13H10N4O2S3
Molecular Weight (g/mol) 350.43
MDL Number MFCD00129148,MFCD00071547
InChI Key COFDRZLHVALCDU-YVLHZVERSA-N
PubChem CID 5702580
IUPAC Name (2Z)-2-(2-amino-1,3-thiazol-4-yl)-1-(1,3-benzothiazol-2-ylsulfanyl)-2-(methoxyimino)ethan-1-one
SMILES CO\N=C(/C(=O)SC1=NC2=CC=CC=C2S1)C1=CSC(N)=N1

Specifications

Melting Point 125°C
Color Yellow
Quantity 5 g
Formula Weight 350.43
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material S-(2-Benzothiazolyl) (Z)-2-(2-Amino-4-thiazolyl)-2-(methoxyimino)thioacetate
RTECSNumber XJ4664500
TSCA No