accessibility menu, dialog, popup

UserName

CAS RN 80756-85-0

IUPAC Name: (2Z)-2-(2-amino-1,3-thiazol-4-yl)-1-(1,3-benzothiazol-2-ylsulfanyl)-2-(methoxyimino)ethan-1-one
Synonyms: (2Z)-2-(2-amino-1,3-thiazol-4-yl)-1-(1,3-benzothiazol-2-ylsulfanyl)-2-(methoxyimino)ethanone
Molecular Weight (g/mol): 350.43
Molecular Formula: C13H10N4O2S3
InChi Key: COFDRZLHVALCDU-YVLHZVERSA-N
SMILES: CO\N=C(/C(=O)SC1=NC2=CC=CC=C2S1)C1=CSC(N)=N1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results